Learn More
Cayman Chemical Aminoethyl-SS-Propionic Acid, Purity: ≥95%, CAS: 15579-00-7, 10 mg
SDP
Catalog No. 502913631
Encompass_Preferred
A cleavable disulfide linker; has been used in the synthesis of ADCs
Catalog No. 50-291-3631 Supplier Cayman Chemical Supplier No. 3550110 MG
A cleavable disulfide linker; has been used in the synthesis of ADCs