Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoferrocene, ≥98.0% (T); 98.0-102.0% (NT), MilliporeSigma™ Supelco™

Catalog No. 111012894 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2894 250 mg
11-101-2893 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2894 Supplier MilliporeSigma Supelco Supplier No. 07379250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD03844760
Synonym eta5-(1-Aminocyclopentadienyl)(cyclopentadienyl)iron(II); Fc-NH2; Ferrocenylamine

Specifications

CAS 1273-82-1
Linear Formula C10H11 FeN
Quantity 250 mg
Formula Weight 201.05
Percent Purity ≥98.0% (T); 98.0 to 102.0% (NT)
Grade For HPLC derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.