Learn More
TOKU-E Amikacin, 37517-28-5, 1 g
Catalog No. 502551847
Encompass_Preferred
aminoglycoside antibiotic, C22H43N5O13. Useful in susceptibility testing, viral transport media, cell culture
Catalog No. 50-255-1847 Supplier TOKU-E Supplier No. A0021G
aminoglycoside antibiotic, C22H43N5O13. Useful in susceptibility testing, viral transport media, cell culture