Learn More
TOKU-E Hygromycin B, 31282-04-9, 500 mg
Catalog No. 502551789
Encompass_Preferred
aminoglycoside antibiotic, selection antibiotic, C20H37N3O13. Useful in cell culture, plant biology.
Catalog No. 50-255-1789 Supplier TOKU-E Supplier No. H007500MG
aminoglycoside antibiotic, selection antibiotic, C20H37N3O13. Useful in cell culture, plant biology.