Learn More
TOKU-E Gentamicin Sulfate, USP, 1405-41-0, 25 g
Catalog No. 502551798
Encompass_Preferred
aminoglycoside antibiotic, selection antibiotic, C60H125N15O25S. Useful in cell culture, cancer research, viral transport media.
Catalog No. 50-255-1798 Supplier TOKU-E Supplier No. G00625G
aminoglycoside antibiotic, selection antibiotic, C60H125N15O25S. Useful in cell culture, cancer research, viral transport media.