Learn More
Aminoguanidine bicarbonate, 98.5%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 125°C |
| Color | White |
| Ignition Residue | 0.3% max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% |
| Packaging | Plastic drum |
| Linear Formula | H2NNHC(=NH)NH2·H2CO3 |
| Quantity | 5 kg |
| Beilstein | 03,117 |
| Solubility Information | Solubility in water: <5g/L (20°C) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic to aquatic life with long lasting effects. May cause an allergic skin reaction. May damage fertility or the unborn child. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention |
| EINECSNumber | 219-956-7 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.