Learn More
Apexbio Technology LLC Aminoguanidine hydrochloride 1937-19-5 100mg
SDP
Catalog No. 502240432
Encompass_Preferred
Aminoguanidine hydrochloride (CAS 1937-19-5) is a small molecule inhibitor targeting diamine oxidase and nitric oxide synthase It has been shown to attenuate doxorubicin-induced apoptosis in a dose-dependent manner in experimental models In addition to its enzyme inhibitory activities aminoguanidine hydrochloride exhibits antioxidant properties These characteristics make it a widely used tool in research focused on the pathophysiology of diabetic nephropathy and related oxidative stress-mediated conditions
Catalog No. 50-224-0432 Supplier Apexbio Technology LLC Supplier No. B6452100
Aminoguanidine hydrochloride (CAS 1937-19-5) is a small molecule inhibitor targeting diamine oxidase and nitric oxide synthase It has been shown to attenuate doxorubicin-induced apoptosis in a dose-dependent manner in experimental models In addition to its enzyme inhibitory activities aminoguanidine hydrochloride exhibits antioxidant properties These characteristics make it a widely used tool in research focused on the pathophysiology of diabetic nephropathy and related oxidative stress-mediated conditions