Learn More
Medchemexpress LLC Aminoguanidine hemisulfate | 996-19-0 | MFCD00035665 | 99.0% | 246.25 g/mol | C2H14N8O4S | 1 G
SDP
Catalog No. 5000299503
Encompass_Preferred
Aminoguanidine hemisulfate is the hemisulfate salt of aminoguanidine used as a research reagent. It acts as an inhibitor of nitric oxide synthase (NOS) and reactive oxygen species (ROS), and has been reported to abolish ANE-induced ROS production in vitro, making it useful in biochemical and cancer research.
  • Inhibits nitric oxide synthase and reduces ROS production.
  • Reported high purity suitable for research applications (reported 99.0%).
  • Solid, white to pink appearance for easy handling.
  • Available in multiple pack sizes for small- and bulk-scale experiments.
  • Stable under recommended storage conditions to preserve activity.
Catalog No. 50-002-99503 Supplier Medchemexpress LLC Supplier No. HYW0207721G
Aminoguanidine hemisulfate is the hemisulfate salt of aminoguanidine used as a research reagent. It acts as an inhibitor of nitric oxide synthase (NOS) and reactive oxygen species (ROS), and has been reported to abolish ANE-induced ROS production in vitro, making it useful in biochemical and cancer research.
  • Inhibits nitric oxide synthase and reduces ROS production.
  • Reported high purity suitable for research applications (reported 99.0%).
  • Solid, white to pink appearance for easy handling.
  • Available in multiple pack sizes for small- and bulk-scale experiments.
  • Stable under recommended storage conditions to preserve activity.