Learn More
Medchemexpress LLC Aminoguanidine hemisulfate | 996-19-0 | MFCD00035665 | 99.0% | 246.25 g/mol | C2H14N8O4S | 500 MG
SDP
Catalog No. 5000301323
Encompass_Preferred
Aminoguanidine hemisulfate is a research reagent that inhibits nitric oxide synthase (NOS) and reduces reactive oxygen species (ROS) production. It is used in cell-based assays to abolish ANE-induced ROS formation and is employed in cancer and oxidative stress research. The compound is supplied as a solid salt with high reported purity suitable for biochemical experiments.
  • Inhibits nitric oxide synthase and reduces ROS production.
  • Abolishes ANE-induced ROS formation in vitro.
  • Used in cancer and oxidative stress research applications.
  • High reported purity (99.0%), suitable for laboratory assays.
  • Solid form, white to pink, easy to weigh and dissolve.
  • Stable when stored frozen; follow recommended storage conditions.
Catalog No. 50-003-01323 Supplier Medchemexpress LLC Supplier No. HYW020772500MG
Aminoguanidine hemisulfate is a research reagent that inhibits nitric oxide synthase (NOS) and reduces reactive oxygen species (ROS) production. It is used in cell-based assays to abolish ANE-induced ROS formation and is employed in cancer and oxidative stress research. The compound is supplied as a solid salt with high reported purity suitable for biochemical experiments.
  • Inhibits nitric oxide synthase and reduces ROS production.
  • Abolishes ANE-induced ROS formation in vitro.
  • Used in cancer and oxidative stress research applications.
  • High reported purity (99.0%), suitable for laboratory assays.
  • Solid form, white to pink, easy to weigh and dissolve.
  • Stable when stored frozen; follow recommended storage conditions.