Learn More
Selleck Chemical LLC Aminoguanidine hydrochloride S4548-100mg
SDP
Catalog No. 503110433
Encompass_Preferred
Aminoguanidine (Pimagedine Guanyl hydrazine Hydrazinecarboximidamide Imino semicarbazide Monoaminoguanidine) is a diamine oxidase and nitric oxide synthase inhibitor It acts to reduce levels of advanced glycation end products (AGEs) through interacting with 3-deoxyglucosone
Catalog No. 50-311-0433 Supplier Selleck Chemical LLC Supplier No. S4548100MG
Aminoguanidine (Pimagedine Guanyl hydrazine Hydrazinecarboximidamide Imino semicarbazide Monoaminoguanidine) is a diamine oxidase and nitric oxide synthase inhibitor It acts to reduce levels of advanced glycation end products (AGEs) through interacting with 3-deoxyglucosone