Learn More
TARGETMOL CHEMICALS INC Aminoguanidine hydrochloride
SDP
Catalog No. 502880618
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Aminoguanidine hydrochloride (Hydrazinecarboximidamide) is a diamine oxidase and NO synthase inhibitor, used in the treatment of diabetic nephropathy. Purity 100%
Catalog No. 50-288-0618 Supplier TARGETMOL CHEMICALS INC Supplier No. T0358500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Aminoguanidine hydrochloride (Hydrazinecarboximidamide) is a diamine oxidase and NO synthase inhibitor, used in the treatment of diabetic nephropathy. Purity 100%