Learn More
Medchemexpress LLC Aminoguanidine (hydrochloride) | 1937-19-5 | ≥98.0% | 50 MG
SDP
Catalog No. 5000413969
Encompass_Preferred
Aminoguanidine hydrochloride (Pimagedine hydrochloride) is an inhibitor of diamine oxidase and nitric oxide synthase. It has a dose-dependent inhibitory effect on apoptosis induced by Doxorubicin and possesses antioxidant properties. It can be utilized in diabetic nephropathy research.
  • Inhibits diamine oxidase and nitric oxide synthase
  • Has a dose-dependent inhibitory effect on apoptosis induced by Doxorubicin
  • Possesses antioxidant properties
  • Can be used in diabetic nephropathy research
Catalog No. 50-004-13969 Supplier Medchemexpress LLC Supplier No. HYB104150MG
Aminoguanidine hydrochloride (Pimagedine hydrochloride) is an inhibitor of diamine oxidase and nitric oxide synthase. It has a dose-dependent inhibitory effect on apoptosis induced by Doxorubicin and possesses antioxidant properties. It can be utilized in diabetic nephropathy research.
  • Inhibits diamine oxidase and nitric oxide synthase
  • Has a dose-dependent inhibitory effect on apoptosis induced by Doxorubicin
  • Possesses antioxidant properties
  • Can be used in diabetic nephropathy research