Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoguanidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. p-7117259 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A112925G 25 g
A1129100G 100 g
A1129500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A112925G Supplier TCI America Supplier No. A112925G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1937-19-5
Molecular Formula CH7ClN4
Molecular Weight (g/mol) 110.545
MDL Number MFCD00039074
InChI Key UBDZFAGVPPMTIT-UHFFFAOYSA-N
Synonym aminoguanidine hydrochloride, pimagedine hcl, guanylhydrazine hydrochloride, hydrazinecarboximidamide hydrochloride, aminoguanidinehydrochloride, aminoguanidine hcl, pimagedine hydrochloride, aminoguanidine monohydrochloride, hydrazinecarboximidamide, hydrochloride
PubChem CID 2734687
IUPAC Name 2-aminoguanidine;hydrochloride
SMILES C(=NN)(N)N.Cl

Specifications

Melting Point 165°C
Color White
Quantity 25 g
Formula Weight 110.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aminoguanidine Hydrochloride
RTECSNumber ME8430000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.