Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminohexylgeldanamycin | 485395-71-9 | 99.6% | 644.80 | C34H52N4O8 | 5 MG
SDP

Catalog No. 5000202080
Encompass_Preferred

Aminohexylgeldanamycin (AHGDM) is a geldanamycin derivative that functions as a potent heat shock protein 90 (HSP90) inhibitor. It has demonstrated antiangiogenic and antitumor activities and is supplied as a high-purity solid for research use.

  • Potent HSP90 inhibitor with antiangiogenic and antitumor activity.
  • High purity: 99.56%.
  • Molecular formula: C34H52N4O8; molecular weight: 644.80.
  • Appearance: solid, purple to black.
  • Storage: solid at -20°C, protect from light; in solvent -80°C (up to 6 months) or -20°C (up to 1 month), protect from light.
  • Synonym: AHGDM.
  • Pack size: 5 mg.

Catalog No. 50-002-02080 Supplier Medchemexpress LLC Supplier No. HY1335715MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminohexylgeldanamycin (AHGDM) is a geldanamycin derivative that functions as a potent heat shock protein 90 (HSP90) inhibitor. It has demonstrated antiangiogenic and antitumor activities and is supplied as a high-purity solid for research use.

  • Potent HSP90 inhibitor with antiangiogenic and antitumor activity.
  • High purity: 99.56%.
  • Molecular formula: C34H52N4O8; molecular weight: 644.80.
  • Appearance: solid, purple to black.
  • Storage: solid at -20°C, protect from light; in solvent -80°C (up to 6 months) or -20°C (up to 1 month), protect from light.
  • Synonym: AHGDM.
  • Pack size: 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.