Learn More
Medchemexpress LLC Aminohexylgeldanamycin | 485395-71-9 | 99.6% | 644.80 g/mol | C34H52N4O8 | 25 MG
SDP
Catalog No. 5000202646
Encompass_Preferred
Aminohexylgeldanamycin (AHGDM) is a geldanamycin derivative and potent HSP90 inhibitor with reported antiangiogenic and antitumor activities. It is supplied as a purified research chemical for in vitro and preclinical studies.
  • Acts as a potent HSP90 inhibitor.
  • Demonstrates antiangiogenic and antitumor activities.
  • High purity (~99.6%) suitable for research applications.
  • Chemical formula C34H52N4O8, molecular weight 644.80 g/mol.
  • Available in small-mass research quantities, for example 25 mg.
Catalog No. 50-002-02646 Supplier Medchemexpress LLC Supplier No. HY13357125MG
Aminohexylgeldanamycin (AHGDM) is a geldanamycin derivative and potent HSP90 inhibitor with reported antiangiogenic and antitumor activities. It is supplied as a purified research chemical for in vitro and preclinical studies.
  • Acts as a potent HSP90 inhibitor.
  • Demonstrates antiangiogenic and antitumor activities.
  • High purity (~99.6%) suitable for research applications.
  • Chemical formula C34H52N4O8, molecular weight 644.80 g/mol.
  • Available in small-mass research quantities, for example 25 mg.