Learn More
Medchemexpress LLC Aminohexylgeldanamycin | 485395-71-9 | MFCD00224213 | 644.8 g/mol | C34H52N4O8 | 10 MG
SDP
Catalog No. 5000202295
Encompass_Preferred
Aminohexylgeldanamycin is a geldanamycin derivative used in research as a potent HSP90 inhibitor with reported antiangiogenic and antitumor activities. It is identified by CAS 485395-71-9 and has molecular formula C34H52N4O8 and molecular weight 644.8 g/mol.
  • Potent HSP90 inhibitor useful for mechanistic and pharmacology studies.
  • Geldanamycin derivative with reported antiangiogenic and antitumor activity.
  • Molecular formula C34H52N4O8, molecular weight 644.8 g/mol.
  • Identified by CAS number 485395-71-9 and MFCD00224213 for database cross-reference.
  • Typical research quantity supplied as 10 mg solid for laboratory use.
Catalog No. 50-002-02295 Supplier Medchemexpress LLC Supplier No. HY13357110MG
Aminohexylgeldanamycin is a geldanamycin derivative used in research as a potent HSP90 inhibitor with reported antiangiogenic and antitumor activities. It is identified by CAS 485395-71-9 and has molecular formula C34H52N4O8 and molecular weight 644.8 g/mol.
  • Potent HSP90 inhibitor useful for mechanistic and pharmacology studies.
  • Geldanamycin derivative with reported antiangiogenic and antitumor activity.
  • Molecular formula C34H52N4O8, molecular weight 644.8 g/mol.
  • Identified by CAS number 485395-71-9 and MFCD00224213 for database cross-reference.
  • Typical research quantity supplied as 10 mg solid for laboratory use.