Learn More
Medchemexpress LLC Aminoiminomethanesulphinic acid | 1758-73-2 | MFCD00002397 | >=99.0% | 108.12 g·mol⁻¹ | CH4N2O2S | 50 G
SDP
Catalog No. 5000303129
Encompass_Preferred
Aminoiminomethanesulphinic acid (thiourea dioxide) is a small-molecule organosulfur reducing agent used in chemical synthesis, analytical methods, and industrial processes. It is a white crystalline solid with molecular formula CH4N2O2S and molecular weight 108.12 g·mol⁻¹. The material is supplied as an analytical-grade reagent with batch-specific quality documentation suitable for research and analytical use.
  • High purity (≥99.0% by NMR)
  • Acts as a mild reducing agent in organic synthesis
  • Soluble in water for aqueous preparations
  • Provided with batch-specific certificate of analysis
  • Suitable for analytical and research applications
Catalog No. 50-003-03129 Supplier Medchemexpress LLC Supplier No. HYW01591550G
Add to Cart
Add to Cart
Aminoiminomethanesulphinic acid (thiourea dioxide) is a small-molecule organosulfur reducing agent used in chemical synthesis, analytical methods, and industrial processes. It is a white crystalline solid with molecular formula CH4N2O2S and molecular weight 108.12 g·mol⁻¹. The material is supplied as an analytical-grade reagent with batch-specific quality documentation suitable for research and analytical use.
  • High purity (≥99.0% by NMR)
  • Acts as a mild reducing agent in organic synthesis
  • Soluble in water for aqueous preparations
  • Provided with batch-specific certificate of analysis
  • Suitable for analytical and research applications