Learn More
Medchemexpress LLC Aminoiminomethanesulphinic acid (thiourea dioxide) | 1758-73-2 | MFCD00002397 | 99.1% | 108.12 g/mol | CH4N2O2S | 250 G
SDP
Catalog No. 5000302476
Encompass_Preferred
Aminoiminomethanesulphinic acid (thiourea dioxide) is a white to off-white solid reagent used in biochemical assays and life-science research. It is supplied as a powder at high assay purity and is soluble in DMSO when warmed; the material is stable under recommended storage conditions for long-term experimental use.
  • High assay purity (99.06%), suitable for analytical applications.
  • White to off-white powder appearance for easy visual inspection.
  • Soluble in DMSO at 10 mg/mL with ultrasonic warming to 60°C.
  • Long-term storage stability: powder stable at -20°C for up to 3 years.
  • Available in multiple pack sizes to match experimental scale.
Catalog No. 50-003-02476 Supplier Medchemexpress LLC Supplier No. 250GHYW015915
Aminoiminomethanesulphinic acid (thiourea dioxide) is a white to off-white solid reagent used in biochemical assays and life-science research. It is supplied as a powder at high assay purity and is soluble in DMSO when warmed; the material is stable under recommended storage conditions for long-term experimental use.
  • High assay purity (99.06%), suitable for analytical applications.
  • White to off-white powder appearance for easy visual inspection.
  • Soluble in DMSO at 10 mg/mL with ultrasonic warming to 60°C.
  • Long-term storage stability: powder stable at -20°C for up to 3 years.
  • Available in multiple pack sizes to match experimental scale.