Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Aminoisobutyric acid, ∽99%, MP Biomedicals™

Catalog No. ICN15035210 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15035210 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15035210 Supplier MP Biomedicals, Inc Supplier No. 0215035210
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53127-08-5
Molecular Formula C4H9NO2
Synonym 2-Methylalanine

Specifications

Color White
Assay Percent Range ∽99%
Quantity 10 g
Formula Weight 103.1
Physical Form Crystalline Powder
Chemical Name or Material α-Aminoisobutyric acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.