Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

δ-Aminolevulinic Acid Hydrochloride, MP Biomedicals™

Catalog No. MP210024725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210024725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210024725 Supplier MP Biomedicals, Inc Supplier No. 0210024725
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in the synthesis of hemes, vitamin B12 and bacteriochlorophyll. δ-Aminolevulinic Acid Hydrochloride, MP Biomedicals is the universal precursor of tetrapyrroles. It is used in porphyrin biosynthesis studies. It enhances chlorophyll formation and

5-Aminolevulinic acid (5-ALA) is an intermediate in heme biosynthesis and being studied as an inducing reagent for protoporphyrin IX (PPIX) dependent fluorescence diagnosis of metastatic lymph nodes. 5-ALA is used for photodynamic therapy of diseases such as Paget’s disease and HPV infection-associated cervical condylomata acuminate.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9-2-5451
Molecular Formula C5H10ClNO3
Molecular Weight (g/mol) 167.59
InChI Key ZLHFONARZHCSET-UHFFFAOYSA-N
Synonym 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
PubChem CID 123608
IUPAC Name 5-amino-4-oxopentanoic acid;hydrochloride
SMILES C(CC(=O)O)C(=O)CN.Cl

Specifications

Melting Point 150°C to 160°C
Solubility Information Soluble in water (5% v/w-clear, colorless solution) .
Quantity 25 mg
Formula Weight 167.59
Percent Purity ∼98%
Grade Biochemical
Physical Form Powder
Recommended Storage –20°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.