Learn More
Description
Thermo Scientific™ AminoLink Coupling Resin is crosslinked 4% beaded agarose that has been activated with aldehyde groups to enable covalent immobilization of antibodies and other proteins through primary amines.
Highlights:
AminoLink Coupling Resin – aldehyde-activated crosslinked 4% beaded agarose
Ideal for antibodies and other proteins – immobilize molecules via primary amines (-NH2)
Flexible coupling conditions – efficient (>85%) coupling over a wide range of pH (4 to 10) and buffer conditions (PBS or other non-amine buffer with or without organic solvent
Stable, permanent immobilization – coupling reaction results in stable, leak-resistant secondary amine bond between resin and ligand
Better than immobilization to CNBr-activated agarose – bond is more stable and uncharged, resulting in less nonspecific binding in affinity purification procedures
Versatile and reusable – prepared affinity resin is adaptable to column and batch affinity techniques and the resin is reusable for typical applications based on protein binding interactions
Includes:
Kit contains pre-packed AminoLink Columns, coupling buffer, quenching buffer, wash solution, sodium cyanoborohydride solution, and column accessories
Specifications
Specifications
| Description | AminoLink Immobilization Kit, 2mL |
| Content And Storage | Sufficient For: Immobilizing 2 to 20 mg of protein per column • AminoLink Columns, 2mL, 5 columns • AminoLink Coupling Buffer (pH 7.0), 250 mL • Quenching Buffer, 50 mL • Wash Buffer, 240 mL • Sodium Cyanoborohydride Solution, 5M, 0.5 mL • Column Accessories Store at 4°C |
| Product Line | AminoLink |
| Quantity | 5 Column Kit |
| Target | Antibody |
| Type | Immobilization Kit |
| Stationary Phase | Aldehyde |
| Sufficient For | 5 Protein Immobilizations |
| Format | Spin Column |
| Column Type | Affinity, Agarose Resin |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.