Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AminoLink™ Plus Coupling Resin

Catalog No. PI20505 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20505 50 mL
PI20501 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI20505 Supplier Thermo Scientific™ Supplier No. 20505
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Covalently couple antibodies and other proteins via primary amines (-NH2) for affinity purification with these aldehyde-activated agarose beads.

Thermo Scientific™ AminoLink Plus Coupling Resin and Kits use aldehyde-activated agarose beads for high-yield covalent coupling of antibodies (proteins) via primary amines for preparation of affinity purification columns.

The resins and kits use activated beaded agarose and a robust coupling chemistry to immobilize proteins and other ligands through primary amines (-NH2) to the resin. Once an antibody or other ligand is immobilized, the prepared affinity resin can be used for a variety of purification methods involving batch or column chromatography. The resin and linkage are stable in binding and elution conditions typically used in affinity chromatography, enabling prepared resin to be used for at least 10 rounds of affinity purification.

The AminoLink Plus Coupling Reaction involves spontaneous formation of Schiff base bonds between aldehydes (on the support) and amines (on the ligand) and their subsequent stabilization by incubation with a mild reductant (sodium cyanoborohydride). The entire coupling reaction, called reductive amination, occurs in 4 to 6 hours in simple non-amine buffers such as PBS. Coupling efficiency with antibodies and typical proteins is generally greater than 85%, resulting in 1 to 20mg of immobilized protein per milliliter of agarose beads.

Highlights:

  • AminoLink Plus Coupling Resin–aldehyde-activated crosslinked 4% beaded agarose

  • Ideal for antibodies and other proteins– immobilize molecules via primary amines (-NH2)

  • Flexible coupling conditions– efficient ( >85%) coupling over a wide range of pH (4-10) and buffer conditions (PBS or other non-amine buffer with or without organic solvent); regular (PBS, pH 7.2) and enhanced (borate, pH 10) coupling protocols provided

  • Stable, permanent immobilization – coupling reaction results in stable, leak-resistant secondary amine bond between resin and ligand

  • Better than immobilization to CNBr-activated agarose – bond is more stable and uncharged, resulting in less nonspecific binding in affinity purification procedures

  • Versatile and reusable – prepared affinity resin is adaptable to column and batch affinity techniques and the resin is reusable for typical applications based on protein binding interactions

  • Convenient kits and product sizes – choose 1- or 5-column kit containing complete sets of buffers, reagents and versatile spin/drip columns, or select bulk resin; bulk quantities available for manufacturing applications

Includes:

Kits contain AminoLink Plus Columns, neutral and enhanced coupling buffers (Part No. 44894 only), quenching buffer, wash solution, sodium cyanoborohydride solution, and column accessories

TRUSTED_SUSTAINABILITY

Specifications

Formulation Aldehyde-activated 4% beaded agarose, slurried in water with sodium azide
Content And Storage Store at 4°C.
Product Line AminoLink
Quantity 50 mL
Target Antibody, Proteins through Primary Amine
Type Resin
Stationary Phase Aldehyde
Format Bottle
Column Type Affinity, Agarose Resin
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.