Learn More
Medchemexpress LLC HY-112052 50mg Medchemexpress, Aminomalonic acid CAS:1068-84-4 Purity:>98%
SDP
Catalog No. 501969000
Encompass_Preferred
Medchemexpress, HY-112052 50mg Aminomalonic acid CAS:1068-84-4 Aminomalonic acid is an amino endogenous metabolite, acts as a strong inhibitor of L-asparagine synthetase from Leukemia 5178Y/AR (Ki= 0.0023 M) and mouse pancreas (Ki= 0.0015 M) in vitro. Aminomalonic acid is a potential biomarker to discriminate between different stages of melanoma metastasis[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9000 Supplier Medchemexpress LLC Supplier No. HY11205250MG
Medchemexpress, HY-112052 50mg Aminomalonic acid CAS:1068-84-4 Aminomalonic acid is an amino endogenous metabolite, acts as a strong inhibitor of L-asparagine synthetase from Leukemia 5178Y/AR (Ki= 0.0023 M) and mouse pancreas (Ki= 0.0015 M) in vitro. Aminomalonic acid is a potential biomarker to discriminate between different stages of melanoma metastasis[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.