Learn More
Biotium Aminooxy-5(6)-TAMRA
Catalog No. 501964823
Encompass_Preferred
Aminooxy-5(6)-ROX reacts with aldehydes or ketones to form a stable hydrazone compound under mild condition. Similar to ARP, aminooxy-5(6)-TAMRA can be used to label abasic sites in damaged DNA. Properties: Ex/Em (MeOH) = 540/565 nm; Dark red solid soluble in DMF or DMSO; Store at -20°C and protect from moisture and light; C 36 H 39 F 6 N 5 O 10; MW: 815.71
Catalog No. 50-196-4823 Supplier Biotium Supplier No. 90105
Add to Cart
Edge
Add to Cart
Aminooxy-5(6)-ROX reacts with aldehydes or ketones to form a stable hydrazone compound under mild condition. Similar to ARP, aminooxy-5(6)-TAMRA can be used to label abasic sites in damaged DNA. Properties: Ex/Em (MeOH) = 540/565 nm; Dark red solid soluble in DMF or DMSO; Store at -20°C and protect from moisture and light; C 36 H 39 F 6 N 5 O 10; MW: 815.71