Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminooxy-amido-PEG4-propargyl | 2253965-03-4 | 97.0% | 304.34 | C13H24N2O6 | 100 MG
SDP

Catalog No. 5000203668
Encompass_Preferred

Aminooxy-amido-PEG4-propargyl is a PEG4 linker bearing an aminooxy group and a terminal propargyl (alkyne) functionality. It is used as a click-chemistry reagent for copper-catalyzed azide-alkyne cycloaddition and as a non-cleavable spacer in antibody-drug conjugate and bioconjugation chemistry.

  • Molecular weight 304.34.
  • Purity 97.0%.
  • Chemical formula C13H24N2O6.
  • CAS number 2253965-03-4.
  • Available in 100 MG and other pack sizes.
  • Suitable for CuAAC click chemistry and bioconjugation.

Catalog No. 50-002-03668 Supplier Medchemexpress LLC Supplier No. HY133435100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminooxy-amido-PEG4-propargyl is a PEG4 linker bearing an aminooxy group and a terminal propargyl (alkyne) functionality. It is used as a click-chemistry reagent for copper-catalyzed azide-alkyne cycloaddition and as a non-cleavable spacer in antibody-drug conjugate and bioconjugation chemistry.

  • Molecular weight 304.34.
  • Purity 97.0%.
  • Chemical formula C13H24N2O6.
  • CAS number 2253965-03-4.
  • Available in 100 MG and other pack sizes.
  • Suitable for CuAAC click chemistry and bioconjugation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.