Learn More
Medchemexpress LLC Aminooxy-amido-PEG4-propargyl | 2253965-03-4 | 97.0% | 304.34 g/mol | C13H24N2O6 | 250 MG
SDP
Catalog No. 5000203669
Encompass_Preferred
Aminooxy-amido-PEG4-propargyl is a viscous, colorless to light yellow PEG-based ADC linker containing an alkyne (propargyl) functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is used as a click chemistry reagent for bioconjugation and antibody-drug conjugate synthesis and is supplied at laboratory research scales.
  • Non-cleavable 4-unit PEG linker.
  • Contains alkyne handle for CuAAC click reactions.
  • Suitable for bioconjugation and ADC synthesis.
  • High purity (~97.0%) for research use.
  • Available in small-scale laboratory quantities.
Catalog No. 50-002-03669 Supplier Medchemexpress LLC Supplier No. HY133435250MG
Aminooxy-amido-PEG4-propargyl is a viscous, colorless to light yellow PEG-based ADC linker containing an alkyne (propargyl) functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is used as a click chemistry reagent for bioconjugation and antibody-drug conjugate synthesis and is supplied at laboratory research scales.
  • Non-cleavable 4-unit PEG linker.
  • Contains alkyne handle for CuAAC click reactions.
  • Suitable for bioconjugation and ADC synthesis.
  • High purity (~97.0%) for research use.
  • Available in small-scale laboratory quantities.