Learn More
Medchemexpress LLC Aminooxy-PEG1-propargyl | 1895922-69-6 | 95.0% | 115.13 | 1 G
SDP
Catalog No. 5000349167
Encompass_Preferred
Aminooxy-PEG1-propargyl is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
Catalog No. 50-003-49167 Supplier Medchemexpress LLC Supplier No. HY1400561G
Aminooxy-PEG1-propargyl is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions