Learn More
Medchemexpress LLC Aminooxy-PEG1-propargyl | 1895922-69-6 | 95.0% | 115.13 | 250 MG
SDP
Catalog No. 5000349394
Encompass_Preferred
Aminooxy-PEG1-propargyl is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group, and is capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules that contain Azide groups
Catalog No. 50-003-49394 Supplier Medchemexpress LLC Supplier No. 250MGHY140056
Aminooxy-PEG1-propargyl is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group, and is capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules that contain Azide groups