Learn More
Medchemexpress LLC Aminooxy-PEG1-propargyl | 1895922-69-6 | 95.0% | 115.13 | 5 G
SDP
Catalog No. 5000349478
Encompass_Preferred
Aminooxy-PEG1-propargyl is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an alkyne group capable of participating in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. It is intended for laboratory research and substance manufacturing applications.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group for CuAAc reactions.
  • Reacts with azide-containing molecules.
  • Suitable for laboratory research.
Catalog No. 50-003-49478 Supplier Medchemexpress LLC Supplier No. HY1400565G
Aminooxy-PEG1-propargyl is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an alkyne group capable of participating in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. It is intended for laboratory research and substance manufacturing applications.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group for CuAAc reactions.
  • Reacts with azide-containing molecules.
  • Suitable for laboratory research.