Learn More
Medchemexpress LLC Aminooxy-PEG1-propargyl | 1895922-69-6 | MFCD29764324 | 95.0% | 115.13 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140056100MG
Encompass_Preferred
Aminooxy-PEG1-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Utilized in the synthesis of PROTACs
  • Selectively degrades target proteins
Catalog No. 50-003-49201