Learn More
Medchemexpress LLC Aminooxy-PEG2-azide | 1043426-13-6 | ≥96.0% | 190.20 | 25 MG
SDP
Catalog No. 5000438680
Encompass_Preferred
Aminooxy-PEG2-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs and serves as a non-cleavable 2-unit PEG ADC linker for synthesizing antibody-drug conjugates (ADCs). This compound is a click chemistry reagent, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Utilized in the synthesis of PROTACs
  • Serves as a non-cleavable 2-unit PEG ADC linker for antibody-drug conjugates
  • Click chemistry reagent containing an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions
  • Participates in strain-promoted alkyne-azide cycloaddition reactions
Catalog No. 50-004-38680 Supplier Medchemexpress LLC Supplier No. 25MGHY113931
Aminooxy-PEG2-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs and serves as a non-cleavable 2-unit PEG ADC linker for synthesizing antibody-drug conjugates (ADCs). This compound is a click chemistry reagent, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Utilized in the synthesis of PROTACs
  • Serves as a non-cleavable 2-unit PEG ADC linker for antibody-drug conjugates
  • Click chemistry reagent containing an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions
  • Participates in strain-promoted alkyne-azide cycloaddition reactions