Learn More
Medchemexpress LLC Aminooxy-PEG2-azide | 1043426-13-6 | ≥96.0% | 190.20 | 250 MG
SDP
Catalog No. 5000439208
Encompass_Preferred
Aminooxy-PEG2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs and as a non-cleavable 2-unit PEG ADC linker for antibody-drug conjugates. It functions as a click chemistry reagent with an Azide group.
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Utilized in the synthesis of PROTACs.
  • Employed in the synthesis of antibody-drug conjugates (ADCs).
Catalog No. 50-004-39208 Supplier Medchemexpress LLC Supplier No. 250MGHY113931
Aminooxy-PEG2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs and as a non-cleavable 2-unit PEG ADC linker for antibody-drug conjugates. It functions as a click chemistry reagent with an Azide group.
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Utilized in the synthesis of PROTACs.
  • Employed in the synthesis of antibody-drug conjugates (ADCs).