Learn More
TARGETMOL CHEMICALS INC Aminooxy-PEG3-azide 50MG
SDP
Catalog No. 502875986
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Aminooxy-PEG3-azide is an ADC linker belonging to the class of PEGs, as well as three-unit PEGs, which can be used for the synthesis of antibody-coupled reactive molecules (ADCs). Purity 95.32%
Catalog No. 50-287-5986 Supplier TARGETMOL CHEMICALS INC Supplier No. T1426450MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Aminooxy-PEG3-azide is an ADC linker belonging to the class of PEGs, as well as three-unit PEGs, which can be used for the synthesis of antibody-coupled reactive molecules (ADCs). Purity 95.32%