Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminooxy-PEG3-C2-NH-Boc | 2062663-65-2 | C13H28N2O6 | 1 G
SDP

Catalog No. 5000357032
Encompass_Preferred

Aminooxy-PEG3-C2-NH-Boc is a PEG- and Alkyl/ether-based PROTAC linker designed for the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.

  • Suitable for PROTAC synthesis
  • Molecular weight: 308.37
  • Targeted for PROTAC linkers pathway
  • Recommended storage: 4°C, sealed, away from moisture and light (solid); -80°C for 6 months or -20°C for 1 month (in solvent)

Catalog No. 50-003-57032 Supplier Medchemexpress LLC Supplier No. HY1404061G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminooxy-PEG3-C2-NH-Boc is a PEG- and Alkyl/ether-based PROTAC linker designed for the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.

  • Suitable for PROTAC synthesis
  • Molecular weight: 308.37
  • Targeted for PROTAC linkers pathway
  • Recommended storage: 4°C, sealed, away from moisture and light (solid); -80°C for 6 months or -20°C for 1 month (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.