Learn More
Medchemexpress LLC Aminooxy-PEG3-C2-NH-Boc | 2062663-65-2 | C13H28N2O6 | 250 MG
SDP
Catalog No. 5000357228
Encompass_Preferred
Aminooxy-PEG3-C2-NH-Boc is a PEG- and Alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG- and Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57228 Supplier Medchemexpress LLC Supplier No. 250MGHY140406
Aminooxy-PEG3-C2-NH-Boc is a PEG- and Alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG- and Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins