Learn More
Medchemexpress LLC Aminooxy-PEG3-C2-NH-Boc | 2062663-65-2 | C13H28N2O6 | 500 MG
SDP
Catalog No. 5000353063
Encompass_Preferred
Aminooxy-PEG3-C2-NH-Boc is a PEG- and Alkyl/ether-based PROTAC linker designed for the synthesis of PROTACs. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins, by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker.
  • PEG- and Alkyl/ether-based PROTAC linker
  • Suitable for the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
Catalog No. 50-003-53063 Supplier Medchemexpress LLC Supplier No. HY140406500MG
Aminooxy-PEG3-C2-NH-Boc is a PEG- and Alkyl/ether-based PROTAC linker designed for the synthesis of PROTACs. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins, by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker.
  • PEG- and Alkyl/ether-based PROTAC linker
  • Suitable for the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system