Learn More
Medchemexpress LLC Aminooxy-PEG3-propargyl | 1807537-27-4 | 98.0% | 203.24 | 25 MG
SDP
Catalog No. 5000351406
Encompass_Preferred
Aminooxy-PEG3-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group for reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-51406 Supplier Medchemexpress LLC Supplier No. 25MGHY140057
Aminooxy-PEG3-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group for reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)