Learn More
Medchemexpress LLC Aminooxy-PEG5-azide | 1919045-02-5 | 98.3% | 100 MG
SDP
Catalog No. 5000340836
Encompass_Preferred
Aminooxy-PEG5-azide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This click chemistry reagent contains an azide group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Acts as a click chemistry reagent
  • Features an azide group for CuAAc reactions
  • Capable of SPAAC reactions with DBCO or BCN groups
Catalog No. 50-003-40836 Supplier Medchemexpress LLC Supplier No. HY130507100MG
Aminooxy-PEG5-azide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This click chemistry reagent contains an azide group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Acts as a click chemistry reagent
  • Features an azide group for CuAAc reactions
  • Capable of SPAAC reactions with DBCO or BCN groups