Learn More
Medchemexpress LLC HY-107994 500mg Medchemexpress, Aminooxyacetic acid hemihydrochloride CAS:2921-14-4 Purity:>98%
SDP
Catalog No. 501968494
Encompass_Preferred
Medchemexpress, HY-107994 500mg Aminooxyacetic acid hemiHCl CAS:2921-14-4 Aminooxyacetic acid hemihydrochloride is a malate-aspartate shuttle (MAS) inhibitor which also inhibits the GABA degradating enzyme GABA-T. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8494 Supplier Medchemexpress LLC Supplier No. HY107994500MG
Medchemexpress, HY-107994 500mg Aminooxyacetic acid hemiHCl CAS:2921-14-4 Aminooxyacetic acid hemihydrochloride is a malate-aspartate shuttle (MAS) inhibitor which also inhibits the GABA degradating enzyme GABA-T. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.