Learn More
Medchemexpress LLC Aminooxyacetic acid hemihydrochloride | 2921-14-4 | MFCD00012955 | >98.0% | 109.30 g·mol⁻1 | C2H5NO3 · 1/2 HCl | 1 ML
SDP
Catalog No. 5000242665
Encompass_Preferred
Aminooxyacetic acid hemihydrochloride is a small-molecule inhibitor of the malate-aspartate shuttle and the GABA-degrading enzyme GABA transaminase (GABA-T). It is supplied as a high-purity solid and as a ready-to-use solution (10 mM in DMSO, 1 mL) for metabolic and neurotransmitter research applications.
  • Inhibits malate-aspartate shuttle and GABA transaminase (GABA-T).
  • High purity suitable for research (>98.0%).
  • Available as solid and as a 10 mM solution in DMSO (1 mL).
  • Recommended storage conditions provided for solid and solution stability.
  • Designed for studies of metabolism and GABAergic signaling.
Catalog No. 50-002-42665 Supplier Medchemexpress LLC Supplier No. HY10799410MM 1ML
Add to Cart
Add to Cart
Aminooxyacetic acid hemihydrochloride is a small-molecule inhibitor of the malate-aspartate shuttle and the GABA-degrading enzyme GABA transaminase (GABA-T). It is supplied as a high-purity solid and as a ready-to-use solution (10 mM in DMSO, 1 mL) for metabolic and neurotransmitter research applications.
  • Inhibits malate-aspartate shuttle and GABA transaminase (GABA-T).
  • High purity suitable for research (>98.0%).
  • Available as solid and as a 10 mM solution in DMSO (1 mL).
  • Recommended storage conditions provided for solid and solution stability.
  • Designed for studies of metabolism and GABAergic signaling.