Learn More
Cusabio Technology LLC Aminopeptidase N ANPEP
Catalog No. 5000051693
Encompass_Preferred
The Mouse Aminopeptidase N (ANPEP) ELISA Kit is designed to detect and quantify ANPEP levels in serum plasma and tissue homogenates using a sandwich immunoassay format With a detection range of 39 mU/mL to 2500 mU/mL and a sensitivity of 9 7 mU/mL this quantitative assay provides reliable results within 1 to 5 hours The kit requires a sample volume of 50-100 L per test and utilizes absorbance measurement at 450 nm for analysis Ideal for research applications it offers a robust method to study ANPEP expression in various biological samples ensuring precision and reproducibility in experimental workflows
Catalog No. 50-000-51693 Supplier Cusabio Technology LLC Supplier No. CSBEL001827MO
The Mouse Aminopeptidase N (ANPEP) ELISA Kit is designed to detect and quantify ANPEP levels in serum plasma and tissue homogenates using a sandwich immunoassay format With a detection range of 39 mU/mL to 2500 mU/mL and a sensitivity of 9 7 mU/mL this quantitative assay provides reliable results within 1 to 5 hours The kit requires a sample volume of 50-100 L per test and utilizes absorbance measurement at 450 nm for analysis Ideal for research applications it offers a robust method to study ANPEP expression in various biological samples ensuring precision and reproducibility in experimental workflows