Learn More
Apexbio Technology LLC Aminophylline 317-34-0 200mg
SDP
Catalog No. 502240719
Encompass_Preferred
Aminophylline (CAS 317-34-0) is a small molecule known for its dual action as a competitive nonselective phosphodiesterase inhibitor and as a nonselective antagonist of adenosine receptors In vitro studies indicate an IC50 of 0 12 mM for phosphodiesterase inhibition highlighting its role in modulating intracellular cyclic nucleotide levels Through these mechanisms aminophylline impacts multiple cellular signaling pathways making it a valuable tool in research focused on smooth muscle relaxation inflammatory response and cellular signaling processes related to adenosine and cyclic AMP regulation
This item is not returnable. View return policy
Catalog No. 50-224-0719 Supplier Apexbio Technology LLC Supplier No. A4346200
Aminophylline (CAS 317-34-0) is a small molecule known for its dual action as a competitive nonselective phosphodiesterase inhibitor and as a nonselective antagonist of adenosine receptors In vitro studies indicate an IC50 of 0 12 mM for phosphodiesterase inhibition highlighting its role in modulating intracellular cyclic nucleotide levels Through these mechanisms aminophylline impacts multiple cellular signaling pathways making it a valuable tool in research focused on smooth muscle relaxation inflammatory response and cellular signaling processes related to adenosine and cyclic AMP regulation