Learn More
Medchemexpress LLC Aminophylline | 317-34-0 | 99.7% | 25 G
SDP
Catalog No. 5000406168
Encompass_Preferred
Aminophylline is a competitive, non-selective phosphodiesterase (PDE) inhibitor and adenosine receptor antagonist. It offers pulmonary vasodilator and bronchodilator actions, aiding asthma research. This dihydrate, a 2:1 ratio compound of theophylline with ethylenediamine, is less potent and shorter-acting than theophylline. It treats airway obstruction in asthma or COPD and serves off-label as a reversal agent during nuclear stress testing.
  • Competitive PDE inhibitor and adenosine receptor antagonist
  • Exhibits pulmonary vasodilator and bronchodilator actions
  • Useful for asthma research
  • Composed of theophylline with ethylenediamine (2:1 ratio)
  • Less potent and shorter-acting than theophylline
  • Treats airway obstruction in asthma or COPD
  • Used off-label for nuclear stress test reversal
  • Increases heart rate and contractility
Catalog No. 50-004-06168 Supplier Medchemexpress LLC Supplier No. 25GHYB0140
Aminophylline is a competitive, non-selective phosphodiesterase (PDE) inhibitor and adenosine receptor antagonist. It offers pulmonary vasodilator and bronchodilator actions, aiding asthma research. This dihydrate, a 2:1 ratio compound of theophylline with ethylenediamine, is less potent and shorter-acting than theophylline. It treats airway obstruction in asthma or COPD and serves off-label as a reversal agent during nuclear stress testing.
  • Competitive PDE inhibitor and adenosine receptor antagonist
  • Exhibits pulmonary vasodilator and bronchodilator actions
  • Useful for asthma research
  • Composed of theophylline with ethylenediamine (2:1 ratio)
  • Less potent and shorter-acting than theophylline
  • Treats airway obstruction in asthma or COPD
  • Used off-label for nuclear stress test reversal
  • Increases heart rate and contractility