Learn More
Selleck Chemical LLC Aminophylline 25mg 317-34-0 Phyllocontin
SDP
Supplier:  Selleck Chemical LLC S167325MG
Encompass_Preferred
Aminophylline (Phyllocontin) is a competitive nonselective phosphodiesterase inhibitor with an IC50 of 0.12 mM and also a nonselective adenosine receptor antagonist. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-494-1