Learn More
Medchemexpress LLC β-Aminopropionitrile | 151-18-8 | MFCD00014820 | 98.99% | 70.09 | 20 G
SDP
Catalog No. 5000365765
Encompass_Preferred
β-Aminopropionitrile (BAPN) is characterized as a specific, irreversible, and orally active inhibitor of lysyl oxidase (LOX). It functions by targeting the active site of LOX or LOXL isoenzymes.
  • Specific, irreversible, and orally active lysyl oxidase (LOX) inhibitor.
  • Targets the active site of LOX or LOXL isoenzymes.
  • Can be used for research in areas such as Monoamine Oxidase, Endogenous Metabolite, Metabolic Disease, and Cancer.
Catalog No. 50-003-65765 Supplier Medchemexpress LLC Supplier No. 20GHYY1750
β-Aminopropionitrile (BAPN) is characterized as a specific, irreversible, and orally active inhibitor of lysyl oxidase (LOX). It functions by targeting the active site of LOX or LOXL isoenzymes.
  • Specific, irreversible, and orally active lysyl oxidase (LOX) inhibitor.
  • Targets the active site of LOX or LOXL isoenzymes.
  • Can be used for research in areas such as Monoamine Oxidase, Endogenous Metabolite, Metabolic Disease, and Cancer.