Learn More
Electron Microscopy Sciences Aminopropylsilane (APS) Slide
SDP
Catalog No. 5031439
Encompass_Preferred
SuperChip Aminopropylsilane coated substrates are designed for microarrays fabricated with the PCR products as probes. Manufactured from the highest quality, low fluorescence glass, SuperChip features a consistent, superior surface coating to provide a uniform DNA immobilizing surface. The result is the slide that binds an optimal amount of probe while minimizing nonspecific binding of labeled targets assuring high density of reactive groups and eliminating virtually all surface artifacts.
These SuperChip substrates are compatible with commercial microarray spotting and detection system. The SuperChip cleaning and coating process is tightly controlled in a HEPA filtered environment. Quality control includes contact angle and background fluorescence measurement of representative samples.
Catalog No. 50-314-39 Supplier Electron Microscopy Sciences Supplier No. 63475AP
SuperChip Aminopropylsilane coated substrates are designed for microarrays fabricated with the PCR products as probes. Manufactured from the highest quality, low fluorescence glass, SuperChip features a consistent, superior surface coating to provide a uniform DNA immobilizing surface. The result is the slide that binds an optimal amount of probe while minimizing nonspecific binding of labeled targets assuring high density of reactive groups and eliminating virtually all surface artifacts.
These SuperChip substrates are compatible with commercial microarray spotting and detection system. The SuperChip cleaning and coating process is tightly controlled in a HEPA filtered environment. Quality control includes contact angle and background fluorescence measurement of representative samples.