Learn More
TARGETMOL CHEMICALS INC AMINOPTERIN 25MG
SDP
Catalog No. 502576291
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Aminopterin (4-Aminofolic acid) is a synthetic folic acid derivative whose metabolite is a competitive inhibitor of dihydrofolate reductase. purity: 98%
Catalog No. 50-257-6291 Supplier TARGETMOL CHEMICALS INC Supplier No. T773025MG
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Aminopterin (4-Aminofolic acid) is a synthetic folic acid derivative whose metabolite is a competitive inhibitor of dihydrofolate reductase. purity: 98%