Learn More
Medchemexpress LLC Aminopterin (4-aminofolic acid) | 54-62-6 | 98.7% | C19H20N8O5 | 100 MG
SDP
Catalog No. 5000355425
Encompass_Preferred
Aminopterin (4-Aminofolic acid) is a 4-amino derivative of folic acid and acts as a folic acid antagonist. It catalyzes the reduction of folic acid to tetrahydrofolic acid and competitively inhibits dihydrofolate reductase (DHFR) with a Ki of 3.7 pM. This compound exhibits anticancer and immunosuppressive activities and is utilized in the treatment of pediatric leukemia.
  • Folic acid antagonist
  • Catalyzes reduction of folic acid to tetrahydrofolic acid
  • Competitively inhibits dihydrofolate reductase (DHFR)
  • Anticancer activity
  • Immunosuppressive activity
  • Used in the treatment of pediatric leukemia
Catalog No. 50-003-55425 Supplier Medchemexpress LLC Supplier No. HY14518100MG
Aminopterin (4-Aminofolic acid) is a 4-amino derivative of folic acid and acts as a folic acid antagonist. It catalyzes the reduction of folic acid to tetrahydrofolic acid and competitively inhibits dihydrofolate reductase (DHFR) with a Ki of 3.7 pM. This compound exhibits anticancer and immunosuppressive activities and is utilized in the treatment of pediatric leukemia.
  • Folic acid antagonist
  • Catalyzes reduction of folic acid to tetrahydrofolic acid
  • Competitively inhibits dihydrofolate reductase (DHFR)
  • Anticancer activity
  • Immunosuppressive activity
  • Used in the treatment of pediatric leukemia