Learn More
Medchemexpress LLC Aminopterin | 54-62-6 | 98.7% | C19H20N8O5 | 50 MG
SDP
Catalog No. 5000353391
Encompass_Preferred
Aminopterin, also known as 4-Aminofolic acid, is a powerful folic acid antagonist. It operates by catalyzing the reduction of folic acid to tetrahydrofolic acid and competitively inhibits dihydrofolate reductase (DHFR) with a high affinity (Ki of 3.7 pM). This compound is recognized for its significant anticancer and immunosuppressive activities, making it a valuable agent in medical applications, particularly in the treatment of pediatric leukemia.
  • Acts as a folic acid antagonist
  • Competitively inhibits dihydrofolate reductase (DHFR)
  • Exhibits anticancer properties
  • Possesses immunosuppressive activities
  • Used in the treatment of pediatric leukemia
Catalog No. 50-003-53391 Supplier Medchemexpress LLC Supplier No. HY1451850MG
Aminopterin, also known as 4-Aminofolic acid, is a powerful folic acid antagonist. It operates by catalyzing the reduction of folic acid to tetrahydrofolic acid and competitively inhibits dihydrofolate reductase (DHFR) with a high affinity (Ki of 3.7 pM). This compound is recognized for its significant anticancer and immunosuppressive activities, making it a valuable agent in medical applications, particularly in the treatment of pediatric leukemia.
  • Acts as a folic acid antagonist
  • Competitively inhibits dihydrofolate reductase (DHFR)
  • Exhibits anticancer properties
  • Possesses immunosuppressive activities
  • Used in the treatment of pediatric leukemia