Learn More
Medchemexpress LLC Aminopterin | 54-62-6 | C19H20N8O5 | 25 MG
SDP
Catalog No. 5000356223
Encompass_Preferred
Aminopterin, also known as 4-Aminofolic acid, is a folic acid antagonist and a 4-amino derivative of folic acid. It functions by catalyzing the reduction of folic acid to tetrahydrofolic acid and competitively inhibiting dihydrofolate reductase (DHFR) with a Kᵢ of 3.7 pM. This compound exhibits both anticancer and immunosuppressive activities, making it useful in the treatment of pediatric leukemia.
  • Folic acid antagonist
  • Inhibits dihydrofolate reductase (DHFR)
  • Exhibits anticancer activity
  • Exhibits immunosuppressive activity
  • Used for pediatric leukemia treatment
  • IC50 value of 4.4 nM against CCRF-CEM cells
Catalog No. 50-003-56223 Supplier Medchemexpress LLC Supplier No. 25MGHY14518
Aminopterin, also known as 4-Aminofolic acid, is a folic acid antagonist and a 4-amino derivative of folic acid. It functions by catalyzing the reduction of folic acid to tetrahydrofolic acid and competitively inhibiting dihydrofolate reductase (DHFR) with a Kᵢ of 3.7 pM. This compound exhibits both anticancer and immunosuppressive activities, making it useful in the treatment of pediatric leukemia.
  • Folic acid antagonist
  • Inhibits dihydrofolate reductase (DHFR)
  • Exhibits anticancer activity
  • Exhibits immunosuppressive activity
  • Used for pediatric leukemia treatment
  • IC50 value of 4.4 nM against CCRF-CEM cells